missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-1,12-diaminododecane Hydrochloride 98.0+%, TCI America™

Catalog No. p-7125970
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34721G 1 g
C34725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34721G Supplier TCI America Supplier No. C34721G
Only null left

Chemical Identifiers

CAS 1051420-16-6
Molecular Formula C20H35ClN2O2
Molecular Weight (g/mol) 370.96
MDL Number MFCD08272292
InChI Key YISCQBUBEASQMD-UHFFFAOYSA-N
Synonym N-Cbz-1,12-diaminododecane Hydrochloride, Benzyl N-(12-Aminododecyl)carbamate Hydrochloride, N-(12-Aminododecyl)carbamic Acid Benzyl Ester Hydrochloride
PubChem CID 131865264
IUPAC Name benzyl N-(12-aminododecyl)carbamate hydrochloride
SMILES Cl.NCCCCCCCCCCCCNC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 175°C
Color White
Quantity 1 g
Formula Weight 370.96
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-1,12-diaminododecane Hydrochloride