CAS RN 1051420-16-6
IUPAC Name:
benzyl N-(12-aminododecyl)carbamate hydrochloride
Synonyms:
benzyl N-(12-aminododecyl)carbamate hydrochloride
Molecular Weight (g/mol):
370.96
Molecular Formula:
C20H35ClN2O2
InChi Key:
YISCQBUBEASQMD-UHFFFAOYSA-N
SMILES:
Cl.NCCCCCCCCCCCCNC(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
N-Carbobenzoxy-1,12-diaminododecane Hydrochloride 98.0+%, TCI America™
CAS: 1051420-16-6 Molecular Formula: C20H35ClN2O2 Molecular Weight (g/mol): 370.96 MDL Number: MFCD08272292 InChI Key: YISCQBUBEASQMD-UHFFFAOYSA-N Synonym: N-Cbz-1,12-diaminododecane Hydrochloride, Benzyl N-(12-Aminododecyl)carbamate Hydrochloride, N-(12-Aminododecyl)carbamic Acid Benzyl Ester Hydrochloride PubChem CID: 131865264 IUPAC Name: benzyl N-(12-aminododecyl)carbamate hydrochloride SMILES: Cl.NCCCCCCCCCCCCNC(=O)OCC1=CC=CC=C1