missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-1,10-diaminodecane Hydrochloride 98.0+%, TCI America™

Catalog No. C34715G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34715G 5 g
C34711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34715G Supplier TCI America Supplier No. C34715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1051420-13-3
Molecular Formula C18H31ClN2O2
Molecular Weight (g/mol) 342.908
MDL Number MFCD08272290
InChI Key XDIZWAPVTINSPI-UHFFFAOYSA-N
Synonym N-Cbz-1,10-diaminodecane Hydrochloride, Benzyl N-(10-Aminodecyl)carbamate Hydrochloride, N-(10-Aminodecyl)carbamic Acid Benzyl Ester Hydrochloride
PubChem CID 131865263
IUPAC Name benzyl N-(10-aminodecyl)carbamate;hydrochloride
SMILES C1=CC=C(C=C1)COC(=O)NCCCCCCCCCCN.Cl

Specifications

Melting Point 176°C
Color White
Quantity 5 g
Formula Weight 342.91
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-1,10-diaminodecane Hydrochloride