accessibility menu, dialog, popup

UserName

OONHNH2HCl

CAS RN 1051420-13-3

IUPAC Name: benzyl N-(10-aminodecyl)carbamate hydrochloride
Synonyms: benzyl N-(10-aminodecyl)carbamate hydrochloride
Molecular Weight (g/mol): 342.91
Molecular Formula: C18H31ClN2O2
InChi Key: XDIZWAPVTINSPI-UHFFFAOYSA-N
SMILES: Cl.NCCCCCCCCCCNC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results