missing translation for 'onlineSavingsMsg'
Learn More

N-Carbamoylmaleimide, tech. 90%

CAS: 3345-50-4 | C5H4N2O3 | 140.098 g/mol

$79.79 - $261.26

Chemical Identifiers

CAS 3345-50-4
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.098
MDL Number MFCD00014539
InChI Key BNPFHEFZJPVCCE-UHFFFAOYSA-N
Synonym n-carbamoylmaleimide, n-carbamylmaleimide, 2,5-dioxo-3-pyrroline-1-carboxamide, 2,5-dioxo-2,5-dihydro-1h-pyrrole-1-carboxamide, n-carbamoylmaleiimide, 3-pyrrolinecarboxamide, 2,5-dioxo, 3-pyrroline-2,5-dione, 1-carbamoyl, 1h-pyrrole-1-carboxamide, 2,5-dihydro-2,5-dioxo-9ci, 2,5-dihydro-2,5-dioxo-1h-pyrrole-1-carboxamide, 2,5-dioxoazolinecarboxamide
PubChem CID 76868
IUPAC Name 2,5-dioxopyrrole-1-carboxamide
SMILES C1=CC(=O)N(C1=O)C(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1368509
View Documents
Thermo Scientific Chemicals
A1368509
10 g
Each for $79.79
Only null left
 
AAA1368518
View Documents
Thermo Scientific Chemicals
A1368518
50 g
Each for $261.26
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3345-50-4
140.098
BNPFHEFZJPVCCE-UHFFFAOYSA-N
76868
C1=CC(=O)N(C1=O)C(=O)N
C5H4N2O3
MFCD00014539
n-carbamoylmaleimide, n-carbamylmaleimide, 2,5-dioxo-3-pyrroline-1-carboxamide, 2,5-dioxo-2,5-dihydro-1h-pyrrole-1-carboxamide, n-carbamoylmaleiimide, 3-pyrrolinecarboxamide, 2,5-dioxo, 3-pyrroline-2,5-dione, 1-carbamoyl, 1h-pyrrole-1-carboxamide, 2,5-dihydro-2,5-dioxo-9ci, 2,5-dihydro-2,5-dioxo-1h-pyrrole-1-carboxamide, 2,5-dioxoazolinecarboxamide
2,5-dioxopyrrole-1-carboxamide

Specifications

3345-50-4
C5H4N2O3
10 g
n-carbamoylmaleimide, n-carbamylmaleimide, 2,5-dioxo-3-pyrroline-1-carboxamide, 2,5-dioxo-2,5-dihydro-1h-pyrrole-1-carboxamide, n-carbamoylmaleiimide, 3-pyrrolinecarboxamide, 2,5-dioxo, 3-pyrroline-2,5-dione, 1-carbamoyl, 1h-pyrrole-1-carboxamide, 2,5-dihydro-2,5-dioxo-9ci, 2,5-dihydro-2,5-dioxo-1h-pyrrole-1-carboxamide, 2,5-dioxoazolinecarboxamide
C1=CC(=O)N(C1=O)C(=O)N
140.098
140.1
Technical
157°C to 160°C
MFCD00014539
117919
BNPFHEFZJPVCCE-UHFFFAOYSA-N
2,5-dioxopyrrole-1-carboxamide
76868
90%
N-Carbamoylmaleimide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 222-097-0

RTECSNumber : UY7352700

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only