missing translation for 'onlineSavingsMsg'
Learn More

N-Carbamoylmaleimide, tech. 90%

Catalog No. AAA1368509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1368509 10 g
AAA1368518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1368509 Supplier Thermo Scientific Chemicals Supplier No. A1368509

CAS: 3345-50-4 | C5H4N2O3 | 140.098 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3345-50-4
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.098
MDL Number MFCD00014539
InChI Key BNPFHEFZJPVCCE-UHFFFAOYSA-N
Synonym n-carbamoylmaleimide, n-carbamylmaleimide, 2,5-dioxo-3-pyrroline-1-carboxamide, 2,5-dioxo-2,5-dihydro-1h-pyrrole-1-carboxamide, n-carbamoylmaleiimide, 3-pyrrolinecarboxamide, 2,5-dioxo, 3-pyrroline-2,5-dione, 1-carbamoyl, 1h-pyrrole-1-carboxamide, 2,5-dihydro-2,5-dioxo-9ci, 2,5-dihydro-2,5-dioxo-1h-pyrrole-1-carboxamide, 2,5-dioxoazolinecarboxamide
PubChem CID 76868
IUPAC Name 2,5-dioxopyrrole-1-carboxamide
SMILES C1=CC(=O)N(C1=O)C(=O)N

Specifications

Melting Point 157°C to 160°C
Quantity 10 g
Beilstein 117919
Formula Weight 140.1
Percent Purity 90%
Grade Technical
Chemical Name or Material N-Carbamoylmaleimide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-097-0
RTECSNumber UY7352700
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only