missing translation for 'onlineSavingsMsg'
Learn More

N-Carbamoyl-DL-aspartic acid, 98%

Catalog No. AAA1716609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1716609 10 g
AAA1716618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1716609 Supplier Thermo Scientific Chemicals Supplier No. A1716609
Only null left

CAS: 923-37-5 | C5H8N2O5 | 176.128 g/mol

N-Carbamoyl-DL-aspartic acid, is a useful biochemical for proteomics research. It is also used as a carbamate derivative which serves as an intermediate in pyrimidine biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Carbamoyl-DL-aspartic acid, is a useful biochemical for proteomics research. It is also used as a carbamate derivative which serves as an intermediate in pyrimidine biosynthesis.

Solubility
Soluble in hot water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 923-37-5
Molecular Formula C5H8N2O5
Molecular Weight (g/mol) 176.128
MDL Number MFCD00042822
InChI Key HLKXYZVTANABHZ-UHFFFAOYSA-N
Synonym ureidosuccinic acid, 2-ureidosuccinic acid, n-carbamoyl-dl-aspartic acid, n-aminocarbonyl-dl-aspartic acid, dl-aspartic acid, n-aminocarbonyl, 2-carbamoylamino butanedioic acid, n-carbamoylaspartic acid, carbamoyl-dl-aspartic acid, carbamylaspartic acid, 2-ureidobutanedioic acid
PubChem CID 279
ChEBI CHEBI:64850
IUPAC Name 2-(carbamoylamino)butanedioic acid
SMILES C(C(C(=O)O)NC(=O)N)C(=O)O

Specifications

Melting Point ∼175°C (decomposition)
Beilstein 1726861
Solubility Information Soluble in hot water.
Quantity 10 g
Formula Weight 176.13
Percent Purity 98%
Chemical Name or Material N-Carbamoyl-DL-aspartic acid
EINECSNumber 213-096-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only