accessibility menu, dialog, popup

UserName

H2NOHNOHOOHO

CAS RN 923-37-5

IUPAC Name: 2-(carbamoylamino)butanedioic acid
Synonyms: ureidosuccinic acid N-carbamoyl-s-aspartic acid NCD N-carbamoyl-S-aspartic acid
Molecular Weight (g/mol): 176.13
Molecular Formula: C5H8N2O5
InChi Key: HLKXYZVTANABHZ-UHFFFAOYNA-N
SMILES: NC(=O)NC(CC(O)=O)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)