missing translation for 'onlineSavingsMsg'
Learn More

n-Butyrophenone, 99%

Catalog No. AC154411000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154411000 100 mL Glass bottle
AC154415000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC154411000 Supplier Thermo Scientific Chemicals Supplier No. 154411000

CAS: 495-40-9 | C10H12O | 148.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 495-40-9
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.2
InChI Key FFSAXUULYPJSKH-UHFFFAOYSA-N
Synonym butyrophenone, n-butyrophenone, 1-butanone, 1-phenyl, phenyl propyl ketone, 1-phenyl-1-butanone, propyl phenyl ketone, unii-186i11wb5b, 1-phenyl-butan-1-one, propylphenylketone
PubChem CID 10315
IUPAC Name 1-phenylbutan-1-one
SMILES CCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 11.0°C to 13.0°C
Color Colorless to Yellow
Density 0.9880g/mL
Boiling Point 228.0°C to 230.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5185 to 1.5205
Linear Formula C6H5COCH2CH2CH3
Quantity 100 mL
Beilstein 07,313
Specific Gravity 0.988
Formula Weight 148.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Butyrophenone
Show More Show Less
EINECSNumber 207-799-7

RUO – Research Use Only