missing translation for 'onlineSavingsMsg'
Learn More

n-Butyltriphenylphosphonium bromide, 99%

Catalog No. p-80712
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108060050 5 g Glass bottle
AC108061000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108060050 Supplier Thermo Scientific Chemicals Supplier No. 108060050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1779-51-7 | C22H24BrP | 399.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 1779-51-7
Molecular Formula C22H24BrP
Molecular Weight (g/mol) 399.31
MDL Number MFCD00011855
InChI Key IKWKJIWDLVYZIY-UHFFFAOYSA-M
Synonym butyltriphenylphosphonium bromide, n-butyltriphenylphosphonium bromide, 1-butyl triphenylphosphonium bromide, phosphonium, butyltriphenyl-, bromide, butyl triphenyl phosphonium bromide, butyl triphenyl phosphine bromide, n-butyltriphenylphosphonium chloride, butyltriphenylphosphanium bromide, phosphonium, butyltriphenyl-, bromide 1:1, n-butyl triphenylphosphonium bromide
PubChem CID 159628
SMILES [Br-].CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 240.0°C to 243.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3(CH2)3P(C6H5)3Br
Quantity 5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,fairly soluble in lower alcohols,slightly soluble in aromatic hydrocarbons
IUPAC Name butyltriphenylphosphanium bromide
Formula Weight 399.3
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material n-Butyltriphenylphosphonium bromide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fre
EINECSNumber 217-219-4
RTECSNumber TA1855200
TSCA TSCA

RUO – Research Use Only