accessibility menu, dialog, popup

UserName

CAS RN 1779-51-7

IUPAC Name: butyltriphenylphosphanium bromide
Synonyms: butyltriphenylphosphanium bromide
Molecular Weight (g/mol): 399.31
Molecular Formula: C22H24BrP
InChi Key: IKWKJIWDLVYZIY-UHFFFAOYSA-M
SMILES: [Br-].CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)