missing translation for 'onlineSavingsMsg'
Learn More

N-Butylpropionamide 99.0+%, TCI America™

Catalog No. B073825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B073825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B073825G Supplier TCI America Supplier No. B073825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2955-67-1
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD00059416
InChI Key XQZDWKBCGAJXLC-UHFFFAOYSA-N
PubChem CID 221431
IUPAC Name N-butylpropanamide
SMILES CCCCNC(=O)CC

Specifications

Color Yellow
Boiling Point 237°C
Quantity 25 g
Formula Weight 129.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Butylpropionamide