accessibility menu, dialog, popup

UserName

H3CNHOCH3

CAS RN 2955-67-1

IUPAC Name: N-butylpropanamide
Synonyms: N-butylpropanamide
Molecular Weight (g/mol): 129.2
Molecular Formula: C7H15NO
InChi Key: XQZDWKBCGAJXLC-UHFFFAOYSA-N
SMILES: CCCCNC(=O)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results