missing translation for 'onlineSavingsMsg'
Learn More

n-Butylmagnesium chloride, 20 wt% solution in THF/Toluene, AcroSeal™

CAS: 693-04-9 | C4H9ClMg | 116.87 g/mol

$111.82 - $111.82

Chemical Identifiers

CAS 693-04-9
Molecular Formula C4H9ClMg
Molecular Weight (g/mol) 116.87
MDL Number MFCD00000475
InChI Key UUNGXYVLXYGZKH-UHFFFAOYSA-M
Synonym butylmagnesium chloride, magnesium, butylchloro, nbumgcl, bumgcl, butylmagnesiumchloride, n-butylmagnesium chloride, n-butyl-magnesium chloride, butylmagnesium chloride solution, butylmagnesium chloride solution, 2.0 m in diethyl ether, unii-4j83fgw852
PubChem CID 101837
SMILES CCCC[Mg]Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC331681000
SDS View Documents
Thermo Scientific Chemicals
331681000
100 mL AcroSeal™ Glass bottle
Each for $111.82
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

693-04-9
116.87
UUNGXYVLXYGZKH-UHFFFAOYSA-M
101837
C4H9ClMg
MFCD00000475
butylmagnesium chloride, magnesium, butylchloro, nbumgcl, bumgcl, butylmagnesiumchloride, n-butylmagnesium chloride, n-butyl-magnesium chloride, butylmagnesium chloride solution, butylmagnesium chloride solution, 2.0 m in diethyl ether, unii-4j83fgw852
CCCC[Mg]Cl

Specifications

693-04-9,109-99-9,108-88-3
19.0,78.0,78.0
Solution
C4H9ClMg
MFCD00000475
butylmagnesium chloride, magnesium, butylchloro, nbumgcl, bumgcl, butylmagnesiumchloride, n-butylmagnesium chloride, n-butyl-magnesium chloride, butylmagnesium chloride solution, butylmagnesium chloride solution, 2.0 m in diethyl ether, unii-4j83fgw852
CCCC[Mg]Cl
116.87
116.87
AcroSeal™ Glass bottle
1.0000g/mL
Brown to Brown-Green
22.0,81.0,81.0
100 mL
CH3(CH2)3MgCl
−21°C
UUNGXYVLXYGZKH-UHFFFAOYSA-M
magnesium;butane;chloride
101837
19 to 22% (as BUMC)
1
n-Butylmagnesium chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger

EINECSNumber : 211-739-5

Recommended Storage : Product may develop some turbidity or precipitate

SDS
Documents

Documents

RUO – Research Use Only