CAS RN 693-04-9
CAS RN 693-04-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
n-Butylmagnesium chloride, 20 wt% solution in THF/Toluene, AcroSeal™
CAS: 693-04-9 Molecular Formula: C4H9ClMg Molecular Weight (g/mol): 116.87 MDL Number: MFCD00000475 InChI Key: UUNGXYVLXYGZKH-UHFFFAOYSA-M Synonym: butylmagnesium chloride,magnesium, butylchloro,nbumgcl,bumgcl,butylmagnesiumchloride,n-butylmagnesium chloride,n-butyl-magnesium chloride,butylmagnesium chloride solution,butylmagnesium chloride solution, 2.0 m in diethyl ether,unii-4j83fgw852 PubChem CID: 101837 SMILES: CCCC[Mg]Cl