missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butylidenephthalide, (E)+(Z), 95%
CAS: 551-08-6 | C12H12O2 | 188.23 g/mol
$69.21 - $660.22
Chemical Identifiers
| CAS | 551-08-6 |
|---|---|
| Molecular Formula | C12H12O2 |
| Molecular Weight (g/mol) | 188.23 |
| MDL Number | MFCD00047319,MFCD00047319 |
| InChI Key | WMBOCUXXNSOQHM-DHZHZOJOSA-N |
| Synonym | ligusticum lactone, butylidene phthalide, n-butylidene phthalide, 3-butylidene-1 3h-isobenzofuranone, 3-butylidenephthalide, phthalide, 3-butylidene, fema no. 3333, bdph, 3-butylidene phthalide, 1 3h-isobenzofuranone, 3-butylidene |
| PubChem CID | 5352899 |
| IUPAC Name | (3E)-3-butylidene-2-benzofuran-1-one |
| SMILES | CCC\C=C1\OC(=O)C2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1035309
|
Thermo Scientific Chemicals
A1035309 |
10 g |
Each for $69.21
|
|
|||||
|
AAA1035318
|
Thermo Scientific Chemicals
A1035318 |
50 g |
Each for $195.95
|
|
|||||
|
AAA1035330
|
Thermo Scientific Chemicals
A1035330 |
250 g |
Each for $660.22
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 551-08-6 | |
| 188.23 | |
| WMBOCUXXNSOQHM-DHZHZOJOSA-N | |
| 5352899 | |
| CCC\C=C1\OC(=O)C2=CC=CC=C12 |
| C12H12O2 | |
| MFCD00047319,MFCD00047319 | |
| ligusticum lactone, butylidene phthalide, n-butylidene phthalide, 3-butylidene-1 3h-isobenzofuranone, 3-butylidenephthalide, phthalide, 3-butylidene, fema no. 3333, bdph, 3-butylidene phthalide, 1 3h-isobenzofuranone, 3-butylidene | |
| (3E)-3-butylidene-2-benzofuran-1-one |
Specifications
| 551-08-6 | |
| 108°C to 109°C (0.1 mmHg) | |
| C12H12O2 | |
| MFCD00047319,MFCD00047319 | |
| 5145 | |
| WMBOCUXXNSOQHM-DHZHZOJOSA-N | |
| (3E)-3-butylidene-2-benzofuran-1-one | |
| 5352899 | |
| 95% |
| 1.103 | |
| 132°C (269°F) | |
| 1.577 | |
| 10 g | |
| ligusticum lactone, butylidene phthalide, n-butylidene phthalide, 3-butylidene-1 3h-isobenzofuranone, 3-butylidenephthalide, phthalide, 3-butylidene, fema no. 3333, bdph, 3-butylidene phthalide, 1 3h-isobenzofuranone, 3-butylidene | |
| CCC\C=C1\OC(=O)C2=CC=CC=C12 | |
| 188.23 | |
| 188.23 | |
| n-Butylidenephthalide, (E)+(Z) |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 208-991-3
RTECSNumber : TI3692500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only