accessibility menu, dialog, popup

UserName

CH3OO(E)

CAS RN 551-08-6

IUPAC Name: (3E)-3-butylidene-1,3-dihydro-2-benzofuran-1-one
Synonyms: butylidenephthalide
Molecular Weight (g/mol): 188.23
Molecular Formula: C12H12O2
InChi Key: WMBOCUXXNSOQHM-DHZHZOJOSA-N
SMILES: CCC\C=C1\OC(=O)C2=CC=CC=C12
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results