missing translation for 'onlineSavingsMsg'
Learn More

n-Butylidenephthalide, (E)+(Z), 95%

Catalog No. AAA1035309
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1035309 10 g
AAA1035318 50 g
AAA1035330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1035309 Supplier Thermo Scientific Chemicals Supplier No. A1035309

CAS: 551-08-6 | C12H12O2 | 188.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 551-08-6
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.23
MDL Number MFCD00047319,MFCD00047319
InChI Key WMBOCUXXNSOQHM-DHZHZOJOSA-N
Synonym ligusticum lactone, butylidene phthalide, n-butylidene phthalide, 3-butylidene-1 3h-isobenzofuranone, 3-butylidenephthalide, phthalide, 3-butylidene, fema no. 3333, bdph, 3-butylidene phthalide, 1 3h-isobenzofuranone, 3-butylidene
PubChem CID 5352899
IUPAC Name (3E)-3-butylidene-2-benzofuran-1-one
SMILES CCC\C=C1\OC(=O)C2=CC=CC=C12

Specifications

Density 1.103
Boiling Point 108°C to 109°C (0.1 mmHg)
Flash Point 132°C (269°F)
Refractive Index 1.577
Quantity 10 g
Beilstein 5145
Formula Weight 188.23
Percent Purity 95%
Chemical Name or Material n-Butylidenephthalide, (E)+(Z)
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 208-991-3
RTECSNumber TI3692500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only