missing translation for 'onlineSavingsMsg'
Learn More

n-Butylethylmagnesium, 0.9M solution in heptane, AcroSeal™, Thermo Scientific™

Chemical Identifiers

CAS 62202-86-2
Molecular Formula C6H14Mg
Molecular Weight (g/mol) 110.48
MDL Number MFCD00072569
InChI Key YHNWUQFTJNJVNU-UHFFFAOYSA-N
Synonym magnesium, butylethyl, butylethylmagnesium, n-butylethylmagnesium, butyl ethyl magnesium, n-butyl ethyl magnesium, mvecfarlyqaunr-uhfffaoysa-n
PubChem CID 112818
IUPAC Name magnesium;butane;ethane
SMILES C[CH2-].CCC[CH2-].[Mg+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC381181000
SDS View Documents
Thermo Scientific Chemicals
381181000
100mL AcroSeal™ Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC381188000
SDS View Documents
Thermo Scientific Chemicals
381188000
800mL AcroSeal™ Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

62202-86-2
110.48
YHNWUQFTJNJVNU-UHFFFAOYSA-N
112818
C[CH2-].CCC[CH2-].[Mg+2]
C6H14Mg
MFCD00072569
magnesium, butylethyl, butylethylmagnesium, n-butylethylmagnesium, butyl ethyl magnesium, n-butyl ethyl magnesium, mvecfarlyqaunr-uhfffaoysa-n
magnesium;butane;ethane

Specifications

62202-86-2,142-82-5
AcroSeal™ Glass Bottle
CH3CH2Mg(CH2)3CH3
100mL
magnesium, butylethyl, butylethylmagnesium, n-butylethylmagnesium, butyl ethyl magnesium, n-butyl ethyl magnesium, mvecfarlyqaunr-uhfffaoysa-n
YHNWUQFTJNJVNU-UHFFFAOYSA-N
magnesium;butane;ethane
112818
n-Butylethylmagnesium, 0.9M Solution in Heptane
0.7300g/mL
C6H14Mg
MFCD00072569
0.73
Solubility in water: reacts
C[CH2-].CCC[CH2-].[Mg+2]
110.48
110.48
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
In contact with water releases

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 263-454-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only