CAS RN 62202-86-2
IUPAC Name:
butyl(ethyl)magnesium
Synonyms:
butyl(ethyl)magnesium
Molecular Weight (g/mol):
110.48
Molecular Formula:
C6H14Mg
InChi Key:
MVECFARLYQAUNR-UHFFFAOYSA-N
SMILES:
CCCC[Mg]CC
1
–
1
of
1
results
1
Promotions Available
n-Butylethylmagnesium, 0.9M solution in heptane, AcroSeal™
n-Butylethylmagnesium, 0.9M solution in heptane, AcroSeal™
CAS: 62202-86-2 Molecular Formula: C6H14Mg Molecular Weight (g/mol): 110.48 MDL Number: MFCD00072569 InChI Key: MVECFARLYQAUNR-UHFFFAOYSA-N Synonym: magnesium, butylethyl,butylethylmagnesium,n-butylethylmagnesium,butyl ethyl magnesium,n-butyl ethyl magnesium,mvecfarlyqaunr-uhfffaoysa-n PubChem CID: 112818 IUPAC Name: magnesium;butane;ethane SMILES: CCCC[Mg]CC