missing translation for 'onlineSavingsMsg'
Learn More

N-Butylethylenediamine 95.0+%, TCI America™

Catalog No. B149125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B149125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B149125ML Supplier TCI America Supplier No. B149125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19522-69-1
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00025602
InChI Key DFPGBRPWDZFIPP-UHFFFAOYSA-N
Synonym n-butylethylenediamine, 2-butylamino ethylamine, n1-butylethane-1,2-diamine, 2-aminoethyl butyl amine, 1,2-ethanediamine,n1-butyl, 2-butylamino-ethylamine, 2-n-butylaminoethylamine, acmc-1bs5m, n-butyl-1,2-ethanediamine, n-butylethane-1,2-diamine
PubChem CID 519668
IUPAC Name N'-butylethane-1,2-diamine
SMILES CCCCNCCN

Specifications

Color Yellow
Boiling Point 175°C
Quantity 25 mL
UN Number 2734
Formula Weight 116.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Butylethylenediamine