missing translation for 'onlineSavingsMsg'
Learn More

N-Butylbenzenesulfonamide 98.0+%, TCI America™

$37.29 - $96.94

Chemical Identifiers

CAS 3622-84-2
Molecular Formula C10H15NO2S
Molecular Weight (g/mol) 213.30
MDL Number MFCD00025024
InChI Key IPRJXAGUEGOFGG-UHFFFAOYSA-N
Synonym benzenesulfonamide, n-butyl, plastomoll bmb, n-n-butylbenzenesulfonamide, dellatol bbs, n-butyl-benzenesulfonamide, benzenesulfonic acid butyl amide, plasthall bsa, cetamoll bmb, uniplex 214, bm 4 sulfonamide
PubChem CID 19241
ChEBI CHEBI:44237
IUPAC Name N-butylbenzenesulfonamide
SMILES CCCCNS(=O)(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B071625G
View Documents
TCI America
B071625G
25 g
Each for $37.29
Only null left
 
B0716500G
View Documents
TCI America
B0716500G
500 g
Each for $96.94
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3622-84-2
213.30
IPRJXAGUEGOFGG-UHFFFAOYSA-N
19241
N-butylbenzenesulfonamide
C10H15NO2S
MFCD00025024
benzenesulfonamide, n-butyl, plastomoll bmb, n-n-butylbenzenesulfonamide, dellatol bbs, n-butyl-benzenesulfonamide, benzenesulfonic acid butyl amide, plasthall bsa, cetamoll bmb, uniplex 214, bm 4 sulfonamide
CHEBI:44237
CCCCNS(=O)(=O)C1=CC=CC=C1

Specifications

3622-84-2
184°C
MFCD00025024
benzenesulfonamide, n-butyl, plastomoll bmb, n-n-butylbenzenesulfonamide, dellatol bbs, n-butyl-benzenesulfonamide, benzenesulfonic acid butyl amide, plasthall bsa, cetamoll bmb, uniplex 214, bm 4 sulfonamide
CCCCNS(=O)(=O)C1=CC=CC=C1
213.30
CHEBI:44237
≥98.0% (HPLC,N)
N-Butylbenzenesulfonamide
Yellow
C10H15NO2S
25 g
IPRJXAGUEGOFGG-UHFFFAOYSA-N
N-butylbenzenesulfonamide
19241
213.30
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1894&(3)-1917&(3)-1930

RTECSNumber : DB1283000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'