missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl vinyl ether, 98%, stab. with 0.01% KOH

Catalog No. p-7027145
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB23673AE 100 mL
AAB23673AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB23673AE Supplier Thermo Scientific Chemicals Supplier No. B23673AE
Only null left

CAS: 111-34-2 | C6H12O | 100.16 g/mol

Butyl Vinyl Ether is a reagent used in organic chemical reactions and the preparation of pharmaceutical compounds used in anticancer therapy. It is also used in the synthesis of potent inhibitors for PDE5. It is also used in the synthesis of copolymers and as an acrylic/vinyl acetate resin comonomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water 3 g/L (20°C). Soluble in chloroform and methanol.

Notes
Store away from air and oxidizing agents may form peroxides. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Donot distill to dryness. Keep it refrigerated. Check the container pressure periodically to prevent peroxide formation.

Chemical Identifiers

CAS 111-34-2
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009454
InChI Key UZKWTJUDCOPSNM-UHFFFAOYSA-N
Synonym n-butyl vinyl ether, butyl vinyl ether, vinyl butyl ether, butane, 1-ethenyloxy, butoxyethylene, 1-ethenyloxy butane, butoxyethene, ether, butyl vinyl, vinyl n-butyl ether, ethenyl n-butyl ether
PubChem CID 8108
IUPAC Name 1-ethenoxybutane
SMILES CCCCOC=C

Specifications

Melting Point -113°C
Density 0.778
Boiling Point 92°C to 94°C
Flash Point −12°C (10°F)
Refractive Index 1.401
Linear Formula CH3(CH2)3OCH=CH2
Quantity 100 mL
UN Number UN3271
Beilstein 1560217
Solubility Information Soluble in water 3g/L (20°C). Soluble in chloroform and methanol.
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material n-Butyl vinyl ether, stabilized with 0.01% KOH
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 203-860-7
RTECSNumber KV5950000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only