accessibility menu, dialog, popup

UserName

H3COCH2

CAS RN 111-34-2

IUPAC Name: 1-(ethenyloxy)butane
Synonyms: butyl vinyl ether
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: UZKWTJUDCOPSNM-UHFFFAOYSA-N
SMILES: CCCCOC=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)