missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl phenyl ether, 99%

Catalog No. AAL0470314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0470314 25 g
AAL0470322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0470314 Supplier Thermo Scientific Chemicals Supplier No. L0470314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1126-79-0 | C10H14O | 150.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1126-79-0
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00009438
InChI Key YFNONBGXNFCTMM-UHFFFAOYSA-N
Synonym butyl phenyl ether, n-butyl phenyl ether, benzene, butoxy, ether, butyl phenyl, phenyl butyl ether, butoxyphenyl, n-butoxybenzene, butyloxy benzene, n-butylphenylether, butoxy-benzene
PubChem CID 14311
IUPAC Name butoxybenzene
SMILES CCCCOC1=CC=CC=C1

Specifications

Melting Point -19°C
Density 0.938
Boiling Point 210°C to 211°C
Flash Point 82°C (179°F)
Refractive Index 1.497
Quantity 25 g
Beilstein 1635559
Formula Weight 150.22
Percent Purity 99%
Chemical Name or Material n-Butyl phenyl ether
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 214-426-1
RTECSNumber KN5300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only