missing translation for 'onlineSavingsMsg'
Learn More

N-Butyl-p-toluenesulfonamide 97.0+%, TCI America™

Catalog No. B074525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B074525G 25 g
B0745500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B074525G Supplier TCI America Supplier No. B074525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1907-65-9
Molecular Formula C11H17NO2S
Molecular Weight (g/mol) 227.322
MDL Number MFCD00027162
InChI Key RQUXYBHREKXNKT-UHFFFAOYSA-N
Synonym n-butyl-p-toluenesulfonamide, n-n-butyl-4-toluenesulfonamide, n-tosylbutylamine, benzenesulfonamide, n-butyl-4-methyl, n-butyltoluene-4-sulfonamide, p-toluenesulfonamide, n-butyl, n-butyl-4-methyl-benzenesulfonamide, unii-7ct68a89du, n-butyltoluene-4-sulphonamide, acmc-1bqiu
PubChem CID 61285
IUPAC Name N-butyl-4-methylbenzenesulfonamide
SMILES CCCCNS(=O)(=O)C1=CC=C(C=C1)C

Specifications

Melting Point 41°C
Color White-Yellow
Boiling Point 234°C
Quantity 25 g
Formula Weight 227.32
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Butyl-p-toluenesulfonamide
EINECSNumber (3)-1929
TSCA Yes