missing translation for 'onlineSavingsMsg'
Learn More

N-Butyl-N-(4-hydroxybutyl)nitrosamine 90.0+%, TCI America™

Catalog No. p-7118758
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09385G 5 g
B093825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09385G Supplier TCI America Supplier No. B09385G
Only null left

Chemical Identifiers

CAS 3817-11-6
Molecular Formula C8H18N2O2
Molecular Weight (g/mol) 174.24
MDL Number MFCD00059008
InChI Key DIKPQFXYECAYPC-UHFFFAOYSA-N
Synonym N-Butyl-N-butan-4-ol-nitrosamine, N-Butyl-N-nitroso-4-aminobutanol, BBN
PubChem CID 19665
IUPAC Name N-butyl-N-(4-hydroxybutyl)nitrous amide
SMILES CCCCN(CCCCO)N=O

Specifications

Boiling Point 167°C
Quantity 5 g
Formula Weight 174.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Butyl-N-(4-hydroxybutyl)nitrosamine
RTECSNumber EL1225000
TSCA No
Recommended Storage Refrigerator