accessibility menu, dialog, popup

UserName

CAS RN 3817-11-6

IUPAC Name: 4-[butyl(nitroso)amino]butan-1-ol
Synonyms: butylhydroxybutylnitrosamine
Molecular Weight (g/mol): 174.24
Molecular Formula: C8H18N2O2
InChi Key: DIKPQFXYECAYPC-UHFFFAOYSA-N
SMILES: CCCCN(CCCCO)N=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results