missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl methacrylate, 99%, stab. with 15-20 ppm 4-methoxyphenol, Thermo Scientific Chemicals

$69.21 - $234.78

Chemical Identifiers

CAS 97-88-1
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00009444
InChI Key SOGAXMICEFXMKE-UHFFFAOYSA-N
Synonym butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate
PubChem CID 7354
IUPAC Name butyl 2-methylprop-2-enoate
SMILES CCCCOC(=O)C(C)=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA19068AP
View Documents
Thermo Scientific Chemicals
A19068AP
500 mL
Each for $69.21
Only null left
 
AAA190680F
View Documents
Thermo Scientific Chemicals
A190680F
2500 mL
Each for $234.78
Only null left
 
Description

Description

n-Butyl methacrylate is used as a building block of polymer based products that are used in paints and coatings, resins, adhesives, toners and inks, oil additives for dental and medical products. Its polymer polybutyl methacrylate (PBMA) with the sodium salt of partially carboxymethylated guar gum (Na-PCMGG, DS = 0.497) is synthesized using ceric ammonium nitrate as a redox initiator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butyl methacrylate is used as a building block of polymer based products that are used in paints and coatings, resins, adhesives, toners and inks, oil additives for dental and medical products. Its polymer polybutyl methacrylate (PBMA) with the sodium salt of partially carboxymethylated guar gum (Na-PCMGG, DS = 0.497) is synthesized using ceric ammonium nitrate as a redox initiator.

Solubility
Miscible with ethyl ether and ethanol. Immiscible with water.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents. Store in a cool place.
Specifications

Chemical Identifiers

97-88-1
142.20
SOGAXMICEFXMKE-UHFFFAOYSA-N
7354
CCCCOC(=O)C(C)=C
C8H14O2
MFCD00009444
butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate
butyl 2-methylprop-2-enoate

Specifications

97-88-1
0.894
49°C (120°F)
C8H14O2
MFCD00009444
UN2227
butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate
SOGAXMICEFXMKE-UHFFFAOYSA-N
butyl 2-methylprop-2-enoate
7354
99%
-75°C
160°C to 163°C
Fruit-like
1.424
500 mL
773960
Miscible with ethyl ether and ethanol. Immiscible with water.
CCCCOC(=O)C(C)=C
142.20
142.2
n-Butyl methacrylate, stabilized with 15 to 20 ppm 4-methoxyphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H226-H315-H319-H317-H335
Toxic if inhaled.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c

H226-H315-H317-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N-BUTYL METHACRYLATE, STABILIZED

EINECSNumber : 202-615-1

RTECSNumber : OZ3675000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only