Learn More
n-Butyl methacrylate, 99%, stab. with 15-20 ppm 4-methoxyphenol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A190680F
Description
n-Butyl methacrylate is used as a building block of polymer based products that are used in paints and coatings, resins, adhesives, toners and inks, oil additives for dental and medical products. Its polymer polybutyl methacrylate (PBMA) with the sodium salt of partially carboxymethylated guar gum (Na-PCMGG, DS = 0.497) is synthesized using ceric ammonium nitrate as a redox initiator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Butyl methacrylate is used as a building block of polymer based products that are used in paints and coatings, resins, adhesives, toners and inks, oil additives for dental and medical products. Its polymer polybutyl methacrylate (PBMA) with the sodium salt of partially carboxymethylated guar gum (Na-PCMGG, DS = 0.497) is synthesized using ceric ammonium nitrate as a redox initiator.
Solubility
Miscible with ethyl ether and ethanol. Immiscible with water.
Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents. Store in a cool place.
Specifications
| n-Butyl methacrylate | |
| 97-88-1 | |
| 0.894 | |
| 49°C (120°F) | |
| C8H14O2 | |
| MFCD00009444 | |
| UN2227 | |
| butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate | |
| SOGAXMICEFXMKE-UHFFFAOYSA-N | |
| butyl 2-methylprop-2-enoate | |
| 7354 | |
| 99% |
| stabilized with 15 to 20 ppm 4-methoxyphenol | |
| -75°C | |
| 160°C to 163°C | |
| Fruit-like | |
| 1.424 | |
| 2500 mL | |
| 773960 | |
| Miscible with ethyl ether and ethanol. Immiscible with water. | |
| CCCCOC(=O)C(C)=C | |
| 142.20 | |
| 142.2 |
Chemical Identifiers
| 97-88-1 | |
| 142.20 | |
| SOGAXMICEFXMKE-UHFFFAOYSA-N | |
| 7354 | |
| CCCCOC(=O)C(C)=C |
| C8H14O2 | |
| MFCD00009444 | |
| butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate | |
| butyl 2-methylprop-2-enoate |
Safety and Handling
GHS H Statement
H331-H226-H315-H319-H317-H335
Toxic if inhaled.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c
H226-H315-H317-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N-BUTYL METHACRYLATE, STABILIZED
EINECSNumber : 202-615-1
RTECSNumber : OZ3675000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only