missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butyl Alcohol, ACS Reagent (1-Butanol), Reagents
n-Butyl Alcohol, ACS Reagent (1-Butanol), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C11200004C
Specifications
| n-Butyl Alcohol, ACS Reagent (1-Butanol) | |
| 71-36-3 | |
| 118°C | |
| Glass Bottle | |
| 0.81 | |
| 1-Butanol, n-Butyl Alcohol, 1-Butyl Alcohol, Butan-1-ol | |
| CCCCO | |
| 74.12 |
| <88.6°C | |
| Colorless | |
| 4 L | |
| Liquid | |
| C4H10O | |
| LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
| butan-1-ol | |
| ACS |
Chemical Identifiers
| 71-36-3 | |
| 74.12 | |
| 1-Butanol, n-Butyl Alcohol, 1-Butyl Alcohol, Butan-1-ol | |
| CCCCO |
| C4H10O | |
| LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
| butan-1-ol |