missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl Alcohol, ACS Reagent (1-Butanol), Reagents

Catalog No. p-200021065
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338155 1 L
01338154 200 L
01338153 20 L
01338152 4 L
01338151 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01338155 Supplier Labchem Inc Supplier No. C11200001C
Only null left

n-Butyl Alcohol, ACS Reagent (1-Butanol), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-Butanol, n-Butyl Alcohol, 1-Butyl Alcohol, Butan-1-ol
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point <88.6°C
Color Colorless
Boiling Point 118°C
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.81
Grade ACS
Chemical Name or Material n-Butyl Alcohol, ACS Reagent (1-Butanol)