Learn More
n-Butyl acetate, ACS reagent
CAS: 123-86-4 | C6H12O2 | 116.16 g/mol
Supplier: Thermo Scientific Chemicals 434621000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| n-Butyl acetate | |
| -78.0°C | |
| 0.8800g/mL | |
| 27°C | |
| Glass bottle | |
| 1.3930 to 1.3950 | |
| MFCD00009445 | |
| 0.001% max. | |
| 15,1537 | |
| Solubility in water: 7g/L (20°C) | |
| CCCCOC(C)=O | |
| 116.16 | |
| 0.69 mPa.s (25°C) | |
| 116.16 | |
| ACS Reagent | |
| Liquid |
| 123-86-4 | |
| Colorless | |
| 127.0°C | |
| Authentic | |
| C6H12O2 | |
| CH3CO2(CH2)3CH3 | |
| 100 mL | |
| 0.88 | |
| n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
| DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
| butyl acetate | |
| 31272 | |
| CHEBI:31328 | |
| ≥99.50%, 0.2% max. (n-Butyl alcohol) | |
| 0.0016 meq/g max |
Chemical Identifiers
| 123-86-4 | |
| 116.16 | |
| DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
| 31272 | |
| butyl acetate |
| C6H12O2 | |
| MFCD00009445 | |
| n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
| CHEBI:31328 | |
| CCCCOC(C)=O |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Avoid breathing dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Warning
EINECSNumber : 204-658-1
RUO – Research Use Only