missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl acetate, ACS reagent

Catalog No. p-4229130
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434621000 100 mL Glass bottle
AC434620010 1 L Glass bottle
AC434620025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC434621000 Supplier Thermo Scientific Chemicals Supplier No. 434621000
Only null left

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Melting Point -78.0°C
Color Colorless
Density 0.8800g/mL
Boiling Point 127.0°C
Flash Point 27°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3930 to 1.3950
Linear Formula CH3CO2(CH2)3CH3
Quantity 100 mL
Residue after Evaporation 0.001% max.
Merck Index 15,1537
Specific Gravity 0.88
Solubility Information Solubility in water: 7g/L (20°C)
Viscosity 0.69 mPa.s (25°C)
Formula Weight 116.16
Percent Purity ≥99.50%, 0.2% max. (n-Butyl alcohol)
Grade ACS Reagent
Titratable Acid 0.0016 meq/g max
Physical Form Liquid
Chemical Name or Material n-Butyl acetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Avoid breathing dust/fume/gas/mist/vapors/spray.
EINECSNumber 204-658-1

RUO – Research Use Only