missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl acetate, ACS reagent

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol

Supplier:  Thermo Scientific Chemicals 434620025

 View more versions of this product

Catalog No. AC434620025

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Butyl acetate
-78.0°C
0.8800g/mL
27°C
Glass bottle
1.3930 to 1.3950
MFCD00009445
0.001% max.
15,1537
Solubility in water: 7g/L (20°C)
CCCCOC(C)=O
116.16
0.69 mPa.s (25°C)
116.16
ACS Reagent
Liquid
123-86-4
Colorless
127.0°C
Authentic
C6H12O2
CH3CO2(CH2)3CH3
2.5 L
0.88
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
DKPFZGUDAPQIHT-UHFFFAOYSA-N
butyl acetate
31272
CHEBI:31328
≥99.50%, 0.2% max. (n-Butyl alcohol)
0.0016 meq/g max

Chemical Identifiers

123-86-4
116.16
DKPFZGUDAPQIHT-UHFFFAOYSA-N
31272
butyl acetate
C6H12O2
MFCD00009445
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
CHEBI:31328
CCCCOC(C)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS Signal Word: Warning

EINECSNumber : 204-658-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only