missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Bromosuccinimide 98.0+%, TCI America™
Supplier: TCI America B065625G
Specifications
| N-Bromosuccinimide | |
| 176°C | |
| C4H4BrNO2 | |
| 25 g | |
| n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide | |
| C1CC(=O)N(C1=O)Br | |
| 177.985 | |
| CHEBI:53174 | |
| ≥98.0% (T) |
| 128-08-5 | |
| White-Yellow | |
| MFCD00005510 | |
| 3261 | |
| PCLIMKBDDGJMGD-UHFFFAOYSA-N | |
| 1-bromopyrrolidine-2,5-dione | |
| 67184 | |
| 177.99 | |
| Crystalline Powder |
Chemical Identifiers
| 128-08-5 | |
| 177.985 | |
| PCLIMKBDDGJMGD-UHFFFAOYSA-N | |
| 67184 | |
| 1-bromopyrrolidine-2,5-dione |
| C4H4BrNO2 | |
| MFCD00005510 | |
| n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide | |
| CHEBI:53174 | |
| C1CC(=O)N(C1=O)Br |
Safety and Handling
EINECSNumber : (5)-0121
TSCA : Yes
Recommended Storage : Refrigerator