missing translation for 'onlineSavingsMsg'
Learn More

N-Bromosuccinimide 98.0+%, TCI America™

Catalog No. p-7118595
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B065625G 25 g
B0656100G 100 g
B0656500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B065625G Supplier TCI America Supplier No. B065625G
Only null left

Chemical Identifiers

CAS 128-08-5
Molecular Formula C4H4BrNO2
Molecular Weight (g/mol) 177.985
MDL Number MFCD00005510
InChI Key PCLIMKBDDGJMGD-UHFFFAOYSA-N
Synonym n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide
PubChem CID 67184
ChEBI CHEBI:53174
IUPAC Name 1-bromopyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)Br

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 177.99
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Bromosuccinimide
EINECSNumber (5)-0121
TSCA Yes
Recommended Storage Refrigerator