missing translation for 'onlineSavingsMsg'
Learn More

N-Bromoacetamide 97.0+%, TCI America™

Catalog No. p-7118500
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B05305G 5 g
B053025G 25 g
2 options

Catalog No. B05305G

Supplier: TCI America B05305G

Only null left

Chemical Identifiers

CAS 79-15-2
Molecular Formula C2H4BrNO
Molecular Weight (g/mol) 137.964
MDL Number MFCD00037097
InChI Key VBTQNRFWXBXZQR-UHFFFAOYSA-N
Synonym acetamide, n-bromo, unii-0aq6mwh7zb, ccris 4590, 0aq6mwh7zb, acetobromamide, n-bromo-acetamide, sjyhiabiktp@, acmc-209pgg, n-bromoacetamide, powder, lopac-b-2377
PubChem CID 4353
IUPAC Name N-bromoacetamide
SMILES CC(=O)NBr

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 137.96
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Bromoacetamide
EINECSNumber (2)-0719
TSCA Yes
Recommended Storage Refrigerator