missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-trans-4-hydroxy-L-prolinol, 96%, Thermo Scientific Chemicals

Catalog No. AAH5105006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5105006 5 g
AAH5105003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5105006 Supplier Thermo Scientific Chemicals Supplier No. H5105006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61478-26-0
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.265
MDL Number MFCD02094386
InChI Key UFJNFQNQLMGUTQ-JGVFFNPUSA-N
PubChem CID 11275979
IUPAC Name tert-butyl (2S,4R)-4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(CC1CO)O

Specifications

Quantity 5 g
Synonym n-boc-trans-4-hydroxy-l-prolinol, boc-hyp-ol, 2s,4r-tert-butyl 4-hydroxy-2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s,4r-4-hydroxy-2-hydroxymethyl pyrrolidine-1-carboxylate, boc-trans-4-hydroxy-l-prolinol, 2s,4r-1-tert-butoxycarbonyl-4-hydroxy-2-hydroxymethyl pyrrolidine, 2s,4r-1-boc-4-hydroxy-2-hydroxymethyl pyrrolidine, n-boc-2s,4r-4-hydroxy-2-hydroxymethylpyrrolidine, 2s,4r-tert-butyl4-hydroxy-2-hydroxymethyl pyrrolidine-1-carboxylate, trans-4-hydroxy-l-prolinol, n-boc protected
Formula Weight 217.26
Percent Purity 96%
Chemical Name or Material N-Boc-trans-4-hydroxy-L-prolinol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only