accessibility menu, dialog, popup

UserName

CAS RN 61478-26-0

IUPAC Name: tert-butyl 4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate
Synonyms: tert-butyl 4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate
Molecular Weight (g/mol): 217.27
Molecular Formula: C10H19NO4
InChi Key: UFJNFQNQLMGUTQ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC(O)CC1CO
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)