missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-trans-4-hydroxy-L-proline benzyl ester, 95%
CAS: 89813-47-8 | C17H23NO5 | 321.373 g/mol
Chemical Identifiers
| CAS | 89813-47-8 |
|---|---|
| Molecular Formula | C17H23NO5 |
| Molecular Weight (g/mol) | 321.373 |
| MDL Number | MFCD00076979 |
| InChI Key | BEIPCYKSYYZEJH-KGLIPLIRSA-N |
| Synonym | boc-hyp-obzl, boc-o-benzyl-l-hydroxyproline, 2s,4r-2-benzyl 1-tert-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 2-benzyl 1-tert-butyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, bochypobn, boc-hyp-obn, 2s,4r-2-benzyl 1-t-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate |
| PubChem CID | 11902907 |
| IUPAC Name | 2-O-benzyl 1-O-tert-butyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate |
| SMILES | CC(C)(C)OC(=O)N1CC(CC1C(=O)OCC2=CC=CC=C2)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH6249503
|
N/A | 1 g |
N/A
|
|
|||||
|
AAH6249506
|
N/A | 5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 89813-47-8 | |
| 321.373 | |
| BEIPCYKSYYZEJH-KGLIPLIRSA-N | |
| 11902907 | |
| CC(C)(C)OC(=O)N1CC(CC1C(=O)OCC2=CC=CC=C2)O |
| C17H23NO5 | |
| MFCD00076979 | |
| boc-hyp-obzl, boc-o-benzyl-l-hydroxyproline, 2s,4r-2-benzyl 1-tert-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 2-benzyl 1-tert-butyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, bochypobn, boc-hyp-obn, 2s,4r-2-benzyl 1-t-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate | |
| 2-O-benzyl 1-O-tert-butyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate |
Specifications
| 89813-47-8 | |
| C17H23NO5 | |
| boc-hyp-obzl, boc-o-benzyl-l-hydroxyproline, 2s,4r-2-benzyl 1-tert-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 2-benzyl 1-tert-butyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, bochypobn, boc-hyp-obn, 2s,4r-2-benzyl 1-t-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate | |
| CC(C)(C)OC(=O)N1CC(CC1C(=O)OCC2=CC=CC=C2)O | |
| 321.373 | |
| 321.37 | |
| Liquid |
| Brown | |
| MFCD00076979 | |
| BEIPCYKSYYZEJH-KGLIPLIRSA-N | |
| 2-O-benzyl 1-O-tert-butyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate | |
| 11902907 | |
| 95% | |
| N-Boc-trans-4-hydroxy-L-proline benzyl ester |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only