missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-sarcosine, 99%

Catalog No. AAL1348206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1348206 5 g
AAL1348214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1348206 Supplier Thermo Scientific Chemicals Supplier No. L1348206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13734-36-6 | C8H15NO4 | 189.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13734-36-6
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.211
MDL Number MFCD00037795
InChI Key YRXIMPFOTQVOHG-UHFFFAOYSA-N
Synonym boc-sar-oh, boc-sarcosine, n-boc-sarcosine, glycine, n-1,1-dimethylethoxy carbonyl-n-methyl, n-tert-butoxycarbonyl-n-methylglycine, t-boc-sarcosine, n-t-boc-sarcosine, 2-tert-butoxy carbonyl methyl amino acetic acid, n-tert-butoxycarbonylsarcosine, tert-butoxycarbonyl methyl amino acetic acid
PubChem CID 83692
IUPAC Name 2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid
SMILES CC(C)(C)OC(=O)N(C)CC(=O)O

Specifications

Melting Point 88°C to 93°C
Beilstein 2046827
Quantity 5 g
Formula Weight 189.2
Percent Purity 99%
Chemical Name or Material N-Boc-sarcosine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-306-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only