accessibility menu, dialog, popup

UserName

CH3NOOOOCH3CH3H3C

CAS RN 13734-36-6

IUPAC Name: 2-{[(tert-butoxy)carbonyl](methyl)amino}acetate
Synonyms: 2-[(tert-butoxycarbonyl)(methyl)amino]acetate
Molecular Weight (g/mol): 188.2
Molecular Formula: C8H14NO4
InChi Key: YRXIMPFOTQVOHG-UHFFFAOYSA-M
SMILES: CN(CC([O-])=O)C(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)