missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-S-trityl-D-cysteine, 98%
CAS: 87494-13-1 | C27H29NO4S | 463.59 g/mol
Supplier: Thermo Scientific Chemicals H6286306
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Boc-S-trityl-D-cysteine | |
| C27H29NO4S | |
| 5 g | |
| JDTOWOURWBDELG-UHFFFAOYNA-N | |
| 2-{[(tert-butoxy)carbonyl]amino}-3-[(triphenylmethyl)sulfanyl]propanoic acid | |
| 11590774 | |
| 98% |
| 87494-13-1 | |
| MFCD00236839 | |
| boc-d-cys trt-oh, boc-s-trityl-d-cysteine, n-alpha-t-butyloxycarbonyl-s-trityl-d-cysteine, s-2-tert-butoxycarbonyl amino-3-tritylthio propanoic acid, boc-l-cysteine trityl, 2s-2-tert-butoxycarbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, boc-cys trt, boc-cys-trt, ambotzbaa5000, n-boc-s-trityl-d-cysteine | |
| CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O | |
| 463.59 | |
| 463.59 |
Chemical Identifiers
| 87494-13-1 | |
| 463.59 | |
| JDTOWOURWBDELG-UHFFFAOYNA-N | |
| 11590774 |
| C27H29NO4S | |
| MFCD00236839 | |
| boc-d-cys trt-oh, boc-s-trityl-d-cysteine, n-alpha-t-butyloxycarbonyl-s-trityl-d-cysteine, s-2-tert-butoxycarbonyl amino-3-tritylthio propanoic acid, boc-l-cysteine trityl, 2s-2-tert-butoxycarbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, boc-cys trt, boc-cys-trt, ambotzbaa5000, n-boc-s-trityl-d-cysteine | |
| CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only