missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-S-trityl-D-cysteine, 98%

Catalog No. AAH6286303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6286303 1 g
AAH6286306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6286303 Supplier Thermo Scientific Chemicals Supplier No. H6286303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 87494-13-1 | C27H29NO4S | 463.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87494-13-1
Molecular Formula C27H29NO4S
Molecular Weight (g/mol) 463.59
MDL Number MFCD00236839
InChI Key JDTOWOURWBDELG-UHFFFAOYNA-N
Synonym boc-d-cys trt-oh, boc-s-trityl-d-cysteine, n-alpha-t-butyloxycarbonyl-s-trityl-d-cysteine, s-2-tert-butoxycarbonyl amino-3-tritylthio propanoic acid, boc-l-cysteine trityl, 2s-2-tert-butoxycarbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, boc-cys trt, boc-cys-trt, ambotzbaa5000, n-boc-s-trityl-d-cysteine
PubChem CID 11590774
SMILES CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Specifications

Quantity 1 g
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-tritylsulfanylpropanoic acid
Formula Weight 463.59
Percent Purity 98%
Chemical Name or Material N-Boc-S-trityl-D-cysteine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only