missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-S-methyl-L-cysteine, 96%

Catalog No. AAH5021703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5021703 1 g
AAH5021706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5021703 Supplier Thermo Scientific Chemicals Supplier No. H5021703
Only null left

CAS: 16947-80-1 | C9H17NO4S | 235.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16947-80-1
Molecular Formula C9H17NO4S
Molecular Weight (g/mol) 235.30
MDL Number MFCD00076923
InChI Key WFDQTEFRLDDJAM-UHFFFAOYNA-N
Synonym n-boc-s-methyl-l-cysteine, boc-cys me-oh, boc-s-methylcysteine, boc-s-methyl-l-cysteine, 2r-2-tert-butoxycarbonyl amino-3-methylsulfanyl propanoic acid, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-methyl, boccys me oh, cysteine, n-1,1-dimethylethoxy carbonyl-s-methyl, n-boc-s-methyl-l-cys-oh, r-boc-s-methyl cysteine
PubChem CID 7018744
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-methylsulfanylpropanoic acid
SMILES CSCC(NC(=O)OC(C)(C)C)C(O)=O

Specifications

Quantity 1 g
Formula Weight 235.3
Percent Purity 96%
Chemical Name or Material N-Boc-S-methyl-L-cysteine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only