accessibility menu, dialog, popup

UserName

CH3CH3H3COOHNSH3COOH

CAS RN 16947-80-1

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-(methylsulfanyl)propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-(methylsulfanyl)propanoic acid
Molecular Weight (g/mol): 235.3
Molecular Formula: C9H17NO4S
InChi Key: WFDQTEFRLDDJAM-UHFFFAOYNA-N
SMILES: CSCC(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results