Learn More
N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid, 98%
CAS: 51077-16-8 | C9H15NO4S | 233.282 g/mol
Supplier: Thermo Scientific Chemicals L1948806
Description
N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-(R)-(-)-thiazolidine-4-carboxylic acid is used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store at 4°C. Store in a cool, dry conditions in well sealed container. Store away from oxidizing agents.
Specifications
| N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid | |
| ∽133°C (decomposition) | |
| MFCD00077002 | |
| 6686084 | |
| Insoluble in water. | |
| −113° (c=1 in Methanol) | |
| (4R)-3-[(2-methylpropan-2-yl)oxycarbonyl]-1,3-thiazolidine-4-carboxylic acid | |
| 688405 | |
| 98% |
| 51077-16-8 | |
| C9H15NO4S | |
| 5 g | |
| boc-thz-oh, 4r-3-tert-butoxycarbonyl-1,3-thiazolane-4-carboxylic acid, n-boc-r-thiazolidine-4-carboxylic acid, 4r-3-tert-butoxycarbonyl-1,3-thiazolidine-4-carboxylic acid, r-3-tert-butoxycarbonyl thiazolidine-4-carboxylic acid, 4r-3-tert-butoxy carbonyl-1,3-thiazolidine-4-carboxylic acid, 3,4-thiazolidinedicarboxylicacid, 3-1,1-dimethylethyl ester, 4r, boc-thioproline, n-boc-thioproline, --boc-l-thioproline | |
| FJWNZTPXVSWUKF-LURJTMIESA-N | |
| CC(C)(C)OC(=O)N1CSCC1C(=O)O | |
| 233.282 | |
| 233.28 |
Chemical Identifiers
| 51077-16-8 | |
| 233.282 | |
| FJWNZTPXVSWUKF-LURJTMIESA-N | |
| 688405 | |
| CC(C)(C)OC(=O)N1CSCC1C(=O)O |
| C9H15NO4S | |
| MFCD00077002 | |
| boc-thz-oh, 4r-3-tert-butoxycarbonyl-1,3-thiazolane-4-carboxylic acid, n-boc-r-thiazolidine-4-carboxylic acid, 4r-3-tert-butoxycarbonyl-1,3-thiazolidine-4-carboxylic acid, r-3-tert-butoxycarbonyl thiazolidine-4-carboxylic acid, 4r-3-tert-butoxy carbonyl-1,3-thiazolidine-4-carboxylic acid, 3,4-thiazolidinedicarboxylicacid, 3-1,1-dimethylethyl ester, 4r, boc-thioproline, n-boc-thioproline, --boc-l-thioproline | |
| (4R)-3-[(2-methylpropan-2-yl)oxycarbonyl]-1,3-thiazolidine-4-carboxylic acid |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only