missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid, 98%

Catalog No. AAL1948803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1948803 1 g
AAL1948806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1948803 Supplier Thermo Scientific Chemicals Supplier No. L1948803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51077-16-8 | C9H15NO4S | 233.282 g/mol

N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store at 4°C. Store in a cool, dry conditions in well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 51077-16-8
Molecular Formula C9H15NO4S
Molecular Weight (g/mol) 233.282
MDL Number MFCD00077002
InChI Key FJWNZTPXVSWUKF-LURJTMIESA-N
Synonym boc-thz-oh, 4r-3-tert-butoxycarbonyl-1,3-thiazolane-4-carboxylic acid, n-boc-r-thiazolidine-4-carboxylic acid, 4r-3-tert-butoxycarbonyl-1,3-thiazolidine-4-carboxylic acid, r-3-tert-butoxycarbonyl thiazolidine-4-carboxylic acid, 4r-3-tert-butoxy carbonyl-1,3-thiazolidine-4-carboxylic acid, 3,4-thiazolidinedicarboxylicacid, 3-1,1-dimethylethyl ester, 4r, boc-thioproline, n-boc-thioproline, --boc-l-thioproline
PubChem CID 688405
IUPAC Name (4R)-3-[(2-methylpropan-2-yl)oxycarbonyl]-1,3-thiazolidine-4-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CSCC1C(=O)O

Specifications

Melting Point ∽133°C (decomposition)
Quantity 1 g
Beilstein 6686084
Solubility Information Insoluble in water.
Optical Rotation −113° (c=1 in Methanol)
Formula Weight 233.28
Percent Purity 98%
Chemical Name or Material N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only