missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-Pyrrole, 98%

Catalog No. p-4323692
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC438510250 25 g
AC438511000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438510250 Supplier Thermo Scientific Chemicals Supplier No. 438510250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5176-27-2 | C9H13NO2 | 167.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5176-27-2
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.21
InChI Key IZPYBIJFRFWRPR-UHFFFAOYSA-N
Synonym tert-butyl 1h-pyrrole-1-carboxylate, n-boc-pyrrole, n-t-boc-pyrrole, tert-butyl 1-pyrrolecarboxylate, t-butyl 1h-pyrrole-1-carboxylate, 1h-pyrrole-1-carboxylic acid, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl-1h-pyrrole, 1-boc pyrrole, 1-boc-pyrrole, acmc-209kvp
PubChem CID 643494
IUPAC Name tert-butyl pyrrole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C=CC=C1

Specifications

Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 97.0°C (2.3 Torr)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 1
Formula Weight 167.21
Percent Purity 98%
Physical Form Solution
Chemical Name or Material N-BOC-Pyrrole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

RUO – Research Use Only